Product Details

SNP ID
rs4657690
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:167631824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAAGCTAGACTCTGTGCTTGCC[A/G]GCAAAGCTTCATGACTTCTCTAAAG
Phenotype
MIM: 610579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCSD1 PubMed Links

Gene Details

Gene
RCSD1
Gene Name
RCSD domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322923.1 Intron NP_001309852.1
NM_001322924.1 Intron NP_001309853.1
NM_052862.3 Intron NP_443094.3

View Full Product Details