Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040424.2 | 6012 | UTR 3 | NP_001035514.1 | ||
NM_001282934.1 | 6012 | UTR 3 | NP_001269863.1 | ||
NM_022115.4 | 6012 | Intron | NP_071398.3 | ||
XM_006724039.2 | 6012 | UTR 3 | XP_006724102.1 | ||
XM_006724040.2 | 6012 | UTR 3 | XP_006724103.1 | ||
XM_011529674.1 | 6012 | UTR 3 | XP_011527976.1 | ||
XM_011529675.1 | 6012 | UTR 3 | XP_011527977.1 | ||
XM_011529676.2 | 6012 | UTR 3 | XP_011527978.1 | ||
XM_011529677.2 | 6012 | UTR 3 | XP_011527979.1 | ||
XM_011529678.2 | 6012 | UTR 3 | XP_011527980.1 | ||
XM_011529679.2 | 6012 | UTR 3 | XP_011527981.1 | ||
XM_011529681.2 | 6012 | UTR 3 | XP_011527983.1 | ||
XM_011529683.1 | 6012 | UTR 3 | XP_011527985.1 | ||
XM_017028425.1 | 6012 | UTR 3 | XP_016883914.1 | ||
XM_017028426.1 | 6012 | UTR 3 | XP_016883915.1 |