Product Details
- SNP ID
-
rs4462
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35301399 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCCCTGGAAATCGCTTGGCAGTAG[A/G]TCTCATAGGTGCTCATTCTTGGGCA
- Phenotype
-
MIM: 604702
MIM: 604700
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HMGXB4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80067987] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HMGXB4
- Gene Name
- HMG-box containing 4
There are no transcripts associated with this gene.
- Gene
- TOM1
- Gene Name
- target of myb1 membrane trafficking protein
View Full Product Details