Product Details

SNP ID
rs11852105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91971065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGATTAGATTATTTTTCTATGAA[G/T]TTGGGCTAATACAATTCCTTTTTAT
Phenotype
MIM: 604505
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIP11 PubMed Links

Gene Details

Gene
TRIP11
Gene Name
thyroid hormone receptor interactor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321851.1 Intron NP_001308780.1
NM_004239.4 Intron NP_004230.2
XM_017021787.1 Intron XP_016877276.1
XM_017021788.1 Intron XP_016877277.1

View Full Product Details