Product Details

SNP ID
rs14739
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:234607746 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGAGTCATGGGGATCTTGAGCCC[C/T]TCTGTGGATGTGGACAGCCACGGCT
Phenotype
MIM: 615332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP2 PubMed Links

Gene Details

Gene
IRF2BP2
Gene Name
interferon regulatory factor 2 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077397.1 1186 Silent Mutation NP_001070865.1
NM_182972.2 1186 Silent Mutation NP_892017.2

View Full Product Details