Product Details

SNP ID
rs34291110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37071032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCATAGACAAAGCCATGGGTTT[C/G]TGAAACAAGTGTGAAATGAAATGTA
Phenotype
MIM: 614571 MIM: 608667
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5orf42 PubMed Links

Gene Details

Gene
C5orf42
Gene Name
chromosome 5 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023073.3 Intron NP_075561.3
XM_005248345.3 Intron XP_005248402.1
XM_005248346.3 Intron XP_005248403.1
XM_005248347.3 Intron XP_005248404.1
XM_005248349.3 Intron XP_005248406.1
XM_005248350.3 Intron XP_005248407.1
XM_006714491.3 Intron XP_006714554.1
XM_011514085.2 Intron XP_011512387.1
XM_011514086.2 Intron XP_011512388.1
XM_011514087.2 Intron XP_011512389.1
XM_011514088.2 Intron XP_011512390.1
XM_011514089.2 Intron XP_011512391.1
XM_011514090.2 Intron XP_011512392.1
XM_011514092.2 Intron XP_011512394.1
XM_011514094.2 Intron XP_011512396.1
XM_017009760.1 Intron XP_016865249.1
XM_017009761.1 Intron XP_016865250.1
XM_017009762.1 Intron XP_016865251.1
XM_017009763.1 Intron XP_016865252.1
XM_017009764.1 Intron XP_016865253.1
XM_017009765.1 Intron XP_016865254.1
XM_017009766.1 Intron XP_016865255.1
Gene
NIPBL
Gene Name
NIPBL, cohesin loading factor
There are no transcripts associated with this gene.

View Full Product Details