Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300914.1 | Intron | NP_001287843.1 | |||
NM_001313964.1 | Intron | NP_001300893.1 | |||
NM_006145.2 | Intron | NP_006136.1 | |||
XM_006722733.1 | Intron | XP_006722796.1 | |||
XM_006722734.3 | Intron | XP_006722797.1 | |||
XM_011527956.2 | Intron | XP_011526258.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321170.1 | Intron | NP_001308099.1 | |||
NM_138501.5 | Intron | NP_612510.1 | |||
XM_006722945.1 | Intron | XP_006723008.1 | |||
XM_011528442.2 | Intron | XP_011526744.1 |