Product Details
- SNP ID
-
rs9926331
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1090261 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGGGAAAGAGGTGGATGCCGGG[C/G]GACTTGGGAGGCAGCAGGCTTGGGC
- Phenotype
-
MIM: 614147
MIM: 182455
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QTNF8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115357486] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QTNF8
- Gene Name
- C1q and tumor necrosis factor related protein 8
- Gene
- SSTR5
- Gene Name
- somatostatin receptor 5
There are no transcripts associated with this gene.
View Full Product Details