Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000296.3 | 13747 | UTR 3 | NP_000287.3 | ||
NM_001009944.2 | 13747 | UTR 3 | NP_001009944.2 | ||
XM_005255370.3 | 13747 | UTR 3 | XP_005255427.1 | ||
XM_011522528.2 | 13747 | UTR 3 | XP_011520830.1 | ||
XM_011522529.2 | 13747 | UTR 3 | XP_011520831.1 | ||
XM_011522530.2 | 13747 | UTR 3 | XP_011520832.1 | ||
XM_011522532.2 | 13747 | UTR 3 | XP_011520834.1 | ||
XM_011522534.2 | 13747 | UTR 3 | XP_011520836.1 | ||
XM_011522535.2 | 13747 | UTR 3 | XP_011520837.1 | ||
XM_011522537.2 | 13747 | UTR 3 | XP_011520839.1 | ||
XM_017023285.1 | 13747 | UTR 3 | XP_016878774.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000548.4 | 13747 | Intron | NP_000539.2 | ||
NM_001077183.2 | 13747 | Intron | NP_001070651.1 | ||
NM_001114382.2 | 13747 | Intron | NP_001107854.1 | ||
NM_001318827.1 | 13747 | Intron | NP_001305756.1 | ||
NM_001318829.1 | 13747 | Intron | NP_001305758.1 | ||
NM_001318831.1 | 13747 | Intron | NP_001305760.1 | ||
NM_001318832.1 | 13747 | Intron | NP_001305761.1 | ||
XM_005255529.4 | 13747 | Intron | XP_005255586.2 | ||
XM_005255531.4 | 13747 | Intron | XP_005255588.2 | ||
XM_011522636.2 | 13747 | Intron | XP_011520938.1 | ||
XM_011522637.2 | 13747 | Intron | XP_011520939.1 | ||
XM_011522638.2 | 13747 | Intron | XP_011520940.2 | ||
XM_011522639.2 | 13747 | Intron | XP_011520941.1 | ||
XM_011522640.2 | 13747 | Intron | XP_011520942.1 | ||
XM_017023615.1 | 13747 | Intron | XP_016879104.1 | ||
XM_017023616.1 | 13747 | Intron | XP_016879105.1 | ||
XM_017023617.1 | 13747 | Intron | XP_016879106.1 | ||
XM_017023618.1 | 13747 | Intron | XP_016879107.1 |