Product Details

SNP ID
rs2237038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:54659666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTCACGAGGGAGTGTTCGGATTC[C/T]CTGGAGGATTTGATCACCTAGCTTA
Phenotype
MIM: 164920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIT PubMed Links

Gene Details

Gene
KIT
Gene Name
KIT proto-oncogene receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000222.2 Intron NP_000213.1
NM_001093772.1 Intron NP_001087241.1
XM_005265740.1 Intron XP_005265797.1
XM_005265741.1 Intron XP_005265798.1
XM_005265742.2 Intron XP_005265799.1
XM_017008178.1 Intron XP_016863667.1
XM_017008179.1 Intron XP_016863668.1
XM_017008180.1 Intron XP_016863669.1

View Full Product Details