Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000774.4 | 308 | Silent Mutation | CCA,CCG | P32P | NP_000765.2 |
XM_011526551.2 | 308 | Silent Mutation | CCA,CCG | P32P | XP_011524853.1 |
XM_011526552.2 | 308 | Silent Mutation | CCA,CCG | P32P | XP_011524854.1 |
XM_011526553.2 | 308 | Silent Mutation | CCA,CCG | P107P | XP_011524855.1 |
XM_011526554.2 | 308 | Intron | XP_011524856.1 | ||
XM_011526555.2 | 308 | Silent Mutation | CCA,CCG | P107P | XP_011524857.1 |
XM_017026384.1 | 308 | Silent Mutation | CCA,CCG | P107P | XP_016881873.1 |
XM_017026385.1 | 308 | Silent Mutation | CCA,CCG | P107P | XP_016881874.1 |
XM_017026386.1 | 308 | Silent Mutation | CCA,CCG | P107P | XP_016881875.1 |
XM_017026387.1 | 308 | Silent Mutation | CCA,CCG | P107P | XP_016881876.1 |