Product Details
- SNP ID
-
rs28732176
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:32129513 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCAGAGATCAGAAGCTTGAAGGG[C/T]CTCTGGCATCTGACTGGTTGATCCA
- Phenotype
-
MIM: 600984
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATF6B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs373328430] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATF6B
- Gene Name
- activating transcription factor 6 beta
There are no transcripts associated with this gene.
- Gene
- FKBPL
- Gene Name
- FK506 binding protein like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022110.3 |
538 |
Missense Mutation |
ACC,GCC |
T90A |
NP_071393.2 |
View Full Product Details