Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308080.1 | 6368 | Intron | NP_001295009.1 | ||
NM_016144.3 | 6368 | Intron | NP_057228.1 | ||
XM_011543458.2 | 6368 | Intron | XP_011541760.1 | ||
XM_011543460.2 | 6368 | Intron | XP_011541762.1 | ||
XM_017009553.1 | 6368 | Intron | XP_016865042.1 | ||
XM_017009555.1 | 6368 | Intron | XP_016865044.1 | ||
XM_017009556.1 | 6368 | Intron | XP_016865045.1 | ||
XM_017009557.1 | 6368 | Intron | XP_016865046.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300780.1 | 6368 | UTR 3 | NP_001287709.1 | ||
NM_020796.4 | 6368 | UTR 3 | NP_065847.1 | ||
XM_005272042.3 | 6368 | Intron | XP_005272099.2 | ||
XM_006714663.1 | 6368 | UTR 3 | XP_006714726.1 | ||
XM_017009675.1 | 6368 | UTR 3 | XP_016865164.1 | ||
XM_017009676.1 | 6368 | UTR 3 | XP_016865165.1 |