Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199189.1 | Intron | NP_001186118.1 | |||
NM_001239.3 | Intron | NP_001230.1 | |||
XM_005248627.3 | Intron | XP_005248684.1 | |||
XM_005248629.4 | Intron | XP_005248686.1 | |||
XM_011543706.2 | Intron | XP_011542008.1 | |||
XM_017010014.1 | Intron | XP_016865503.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002890.2 | Intron | NP_002881.1 | |||
NM_022650.2 | Intron | NP_072179.1 | |||
XM_011543525.1 | Intron | XP_011541827.1 | |||
XM_011543527.2 | Intron | XP_011541829.1 |