Product Details

SNP ID
rs1803001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71239903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACCTCCAATGCAAGATGGGGAAG[A/G]GAGAAGATGTAGGCAGTGGCGGCAG
Phenotype
MIM: 300061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZMYM3 PubMed Links

Gene Details

Gene
ZMYM3
Gene Name
zinc finger MYM-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171162.1 5314 UTR 3 NP_001164633.1
NM_001171163.1 5314 Intron NP_001164634.1
NM_005096.3 5314 UTR 3 NP_005087.1
NM_201599.2 5314 UTR 3 NP_963893.1
XM_005262309.3 5314 UTR 3 XP_005262366.1
XM_005262310.2 5314 UTR 3 XP_005262367.1
XM_011531062.2 5314 UTR 3 XP_011529364.1

View Full Product Details