Product Details

SNP ID
rs1502683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:82984665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTAACACCCGGGAAGACTCATTT[C/T]AGGTTTCTGACCTCCAAAATTGTCA
Phenotype
MIM: 611344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASEF PubMed Links

Gene Details

Gene
RASEF
Gene Name
RAS and EF-hand domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152573.3 Intron NP_689786.2
XM_005251730.3 Intron XP_005251787.1
XM_005251731.3 Intron XP_005251788.1

View Full Product Details