Product Details
- SNP ID
-
rs1260731
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:125286157 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTCTGTGAGGCATTCATGAGGAA[C/T]CTACTAGGCAACAAGTCTCAGAAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HDDC2
PubMed Links
Gene Details
- Gene
- HDDC2
- Gene Name
- HD domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016063.2 |
|
Intron |
|
|
NP_057147.2 |
View Full Product Details