Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173662.2 | 1402 | Missense Mutation | ATA,ATG | I315M | NP_775933.1 |
XM_005262938.3 | 1402 | Missense Mutation | ATA,ATG | I281M | XP_005262995.1 |
XM_005262939.3 | 1402 | Missense Mutation | ATA,ATG | I235M | XP_005262996.1 |
XM_005262940.4 | 1402 | Missense Mutation | ATA,ATG | I234M | XP_005262997.1 |
XM_011531879.2 | 1402 | Missense Mutation | ATA,ATG | I283M | XP_011530181.1 |
XM_011531881.2 | 1402 | Missense Mutation | ATA,ATG | I275M | XP_011530183.1 |
XM_011531882.2 | 1402 | Intron | XP_011530184.1 | ||
XM_011531883.2 | 1402 | Missense Mutation | ATA,ATG | I243M | XP_011530185.1 |
XM_017008047.1 | 1402 | Missense Mutation | ATA,ATG | I216M | XP_016863536.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318787.1 | 1402 | Intron | NP_001305716.1 | ||
NM_001318789.1 | 1402 | Intron | NP_001305718.1 | ||
NM_001318790.1 | 1402 | Intron | NP_001305719.1 | ||
NM_001318791.1 | 1402 | Intron | NP_001305720.1 | ||
NM_001318793.1 | 1402 | Intron | NP_001305722.1 | ||
NM_001318795.1 | 1402 | Intron | NP_001305724.1 | ||
NM_001318796.1 | 1402 | Intron | NP_001305725.1 | ||
NM_003264.4 | 1402 | Intron | NP_003255.2 | ||
XM_011532215.2 | 1402 | UTR 3 | XP_011530517.1 | ||
XM_011532216.2 | 1402 | UTR 3 | XP_011530518.1 | ||
XM_017008573.1 | 1402 | UTR 3 | XP_016864062.1 | ||
XM_017008574.1 | 1402 | UTR 3 | XP_016864063.1 | ||
XM_017008575.1 | 1402 | UTR 3 | XP_016864064.1 | ||
XM_017008576.1 | 1402 | UTR 3 | XP_016864065.1 |