Product Details

SNP ID
rs708165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27081267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGTTGGTGCTGACCGATGGGGACT[C/T]TTGGTGGCATTCACAGGGTGATCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf71 PubMed Links

Gene Details

Gene
C12orf71
Gene Name
chromosome 12 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080406.1 1298 Silent Mutation AAA,AAG K239K NP_001073875.1
XM_011520847.2 1298 Silent Mutation AAA,AAG K239K XP_011519149.1

View Full Product Details