Product Details

SNP ID
rs1131266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241675983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTGCACCCCAGCTCCGGGGCA[A/G]AAGAGGCAGCCTGCAGATCTCTGCT
Phenotype
MIM: 611338 MIM: 188345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG4B PubMed Links

Gene Details

Gene
ATG4B
Gene Name
autophagy related 4B cysteine peptidase
There are no transcripts associated with this gene.

Gene
DTYMK
Gene Name
deoxythymidylate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165031.1 805 UTR 3 NP_001158503.1
NM_001320902.1 805 UTR 3 NP_001307831.1
NM_001320903.1 805 UTR 3 NP_001307832.1
NM_001320904.1 805 UTR 3 NP_001307833.1
NM_001320905.1 805 UTR 3 NP_001307834.1
NM_012145.3 805 UTR 3 NP_036277.2

View Full Product Details