Product Details

SNP ID
rs6561
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:27301794 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTTATTTTTCCTTTTAGTGAGAT[C/T]ACGTACTATAGGTGAACTTTTAGCT
Phenotype
MIM: 610091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4522 PubMed Links

Gene Details

Gene
MIR4522
Gene Name
microRNA 4522
There are no transcripts associated with this gene.

Gene
WSB1
Gene Name
WD repeat and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015626.8 462 Missense Mutation TCA,TTA S16L NP_056441.6
NM_134265.2 462 Intron NP_599027.1
XM_005257963.3 462 Intron XP_005258020.1
XM_017024433.1 462 Missense Mutation TCA,TTA S16L XP_016879922.1
XM_017024434.1 462 UTR 5 XP_016879923.1
XM_017024435.1 462 UTR 5 XP_016879924.1
XM_017024436.1 462 Intron XP_016879925.1

View Full Product Details