Product Details

SNP ID
rs1062463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17812723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCTTCTCGAGTGTGCGCGCCG[C/G]CTCAGCCAGCATGCCCACGCGCTGC
Phenotype
MIM: 613486 MIM: 607642 MIM: 184756
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR33B PubMed Links

Gene Details

Gene
MIR33B
Gene Name
microRNA 33b
There are no transcripts associated with this gene.

Gene
MIR6777
Gene Name
microRNA 6777
There are no transcripts associated with this gene.

Gene
RAI1
Gene Name
retinoic acid induced 1
There are no transcripts associated with this gene.

Gene
SREBF1
Gene Name
sterol regulatory element binding transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005291.2 3537 Missense Mutation CCG,GCG P1145A NP_001005291.1
NM_001321096.2 3537 Missense Mutation CCG,GCG P1091A NP_001308025.1
NM_004176.4 3537 Missense Mutation CCG,GCG P1115A NP_004167.3
XM_005256772.4 3537 Missense Mutation CCG,GCG P1113A XP_005256829.1
XM_017024970.1 3537 Missense Mutation CCG,GCG P734A XP_016880459.1
XM_017024971.1 3537 Missense Mutation CCG,GCG P634A XP_016880460.1

View Full Product Details