Product Details

SNP ID
rs1421654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73566142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAATCAGCAACTCTCCAAGGCA[A/G]TGGACACATTTCCATCTTTGCCTTT
Phenotype
MIM: 605787 MIM: 616194
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 389 Intron NP_075526.1
XM_005248560.2 389 Intron XP_005248617.1
XM_017009678.1 389 Intron XP_016865167.1
Gene
UTP15
Gene Name
UTP15, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284430.1 389 Intron NP_001271359.1
NM_001284431.1 389 Intron NP_001271360.1
NM_032175.3 389 Intron NP_115551.2
XM_011543680.2 389 UTR 5 XP_011541982.1

View Full Product Details