Product Details

SNP ID
rs7409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76039462 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGCACCAGACAGCAGCGGCCCC[C/T]TTCCCAGGAGGTACAGGAGACAGGA
Phenotype
MIM: 604858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRP68 PubMed Links

Gene Details

Gene
SRP68
Gene Name
signal recognition particle 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260502.1 2163 UTR 3 NP_001247431.1
NM_001260503.1 2163 UTR 3 NP_001247432.1
NM_014230.3 2163 UTR 3 NP_055045.2

View Full Product Details