Product Details

SNP ID
rs1065768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78174749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCAGGAAGGGAGCGGGCGGCCCT[C/T]GCAGGTCCCTCAGTTGGCAGGGCTG
Phenotype
MIM: 603926 MIM: 188300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf99 PubMed Links

Gene Details

Gene
C17orf99
Gene Name
chromosome 17 open reading frame 99
There are no transcripts associated with this gene.

Gene
SYNGR2
Gene Name
synaptogyrin 2
There are no transcripts associated with this gene.

Gene
TK1
Gene Name
thymidine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003258.4 1028 UTR 3 NP_003249.3
XM_017024992.1 1028 UTR 3 XP_016880481.1

View Full Product Details