Product Details
- SNP ID
-
rs1122389
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:123646881 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATAGAAGGTGCCAAGTTTGTGTCA[C/T]GCCCAGTGATCCTCTGTTCAGACTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL38
PubMed Links
Gene Details
- Gene
- KLHL38
- Gene Name
- kelch like family member 38
View Full Product Details