Product Details
- SNP ID
-
rs1356999
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:24127514 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTAGGAAACCAACAGCCTTATAG[A/T]TGGCAATGGCTTTTTAATCAAATTT
- Phenotype
-
MIM: 616630
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NRSN1
PubMed Links
Gene Details
- Gene
- NRSN1
- Gene Name
- neurensin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080723.4 |
|
Intron |
|
|
NP_542454.3 |
View Full Product Details