Product Details

SNP ID
rs13900
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:34256892 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTCCCCTAGCTTTCCCCAGACACC[C/T]TGTTTTATTTTATTATAATGAATTT
Phenotype
MIM: 158105
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL2 PubMed Links

Gene Details

Gene
CCL2
Gene Name
C-C motif chemokine ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002982.3 438 UTR 3 NP_002973.1

View Full Product Details