Product Details

SNP ID
rs1059658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44319853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGAGGAAGTGCTGGGCCGCCCA[A/G]AGGGACAGCCCTGCCCTGGAGCTTG
Phenotype
MIM: 605448 MIM: 610820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RUNDC3A PubMed Links

Gene Details

Gene
RUNDC3A
Gene Name
RUN domain containing 3A
There are no transcripts associated with this gene.

Gene
SLC25A39
Gene Name
solute carrier family 25 member 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143780.2 1318 UTR 3 NP_001137252.1
NM_001321240.1 1318 UTR 3 NP_001308169.1
NM_001321241.1 1318 UTR 3 NP_001308170.1
NM_016016.3 1318 UTR 3 NP_057100.1
XM_011524880.2 1318 UTR 3 XP_011523182.1
XM_011524882.2 1318 UTR 3 XP_011523184.1
XM_011524883.2 1318 UTR 3 XP_011523185.1
XM_011524884.1 1318 UTR 3 XP_011523186.1
XM_017024737.1 1318 UTR 3 XP_016880226.1

View Full Product Details