Product Details

SNP ID
rs1541184
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:19814790 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ATGGCTGGGCCAGCTGCCCCACCAC[C/T]GCCTCATGGTCGCGCAGGCTGCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF186 PubMed Links

Gene Details

Gene
RNF186
Gene Name
ring finger protein 186
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019062.1 489 Silent Mutation GCA,GCG A104A NP_061935.1

View Full Product Details