Product Details

SNP ID
rs927879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110869004 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCAGGACGGAGCCGCACATCTG[C/T]TCTCCTCCACCTTTGGTCAATTCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD10 PubMed Links

Gene Details

Gene
ANKRD10
Gene Name
ankyrin repeat domain 10
There are no transcripts associated with this gene.

Gene
LINC00346
Gene Name
long intergenic non-protein coding RNA 346
There are no transcripts associated with this gene.

View Full Product Details