Product Details

SNP ID
rs1803325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1423185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGGGTAAGCAGTGTGTAACACT[C/T]CTTCCTGTCCATCGGTTTTCCACAG
Phenotype
MIM: 164762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005206.4 1393 UTR 3 NP_005197.3
NM_016823.3 1393 UTR 3 NP_058431.2

View Full Product Details