Product Details

SNP ID
rs2256322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82735829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCGGGTCTCTGCGGGGCCTGGG[A/G]AGGCGGAGGGGTCGGGGGCAGGCGC
Phenotype
MIM: 608425 MIM: 611683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FN3K PubMed Links

Gene Details

Gene
FN3K
Gene Name
fructosamine 3 kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022158.3 Intron NP_071441.1
Gene
FN3KRP
Gene Name
fructosamine 3 kinase related protein
There are no transcripts associated with this gene.

View Full Product Details