Product Details

SNP ID
rs1018364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16225834 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGATCAAACCCAAGAGCATTCA[C/G]TGTGGAATGCTGTGAAACAAATTGT
Phenotype
MIM: 600849 MIM: 605947
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NCOR1 PubMed Links

Gene Details

Gene
NCOR1
Gene Name
nuclear receptor corepressor 1
There are no transcripts associated with this gene.

Gene
PIGL
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004278.3 Intron NP_004269.1
XM_011524080.2 Intron XP_011522382.1
XM_017025349.1 Intron XP_016880838.1
XM_017025350.1 Intron XP_016880839.1
XM_017025351.1 Intron XP_016880840.1
XM_017025352.1 Intron XP_016880841.1
XM_017025353.1 Intron XP_016880842.1
XM_017025354.1 Intron XP_016880843.1
XM_017025355.1 Intron XP_016880844.1
XM_017025356.1 Intron XP_016880845.1

View Full Product Details