Product Details

SNP ID
hCV7460920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55937340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGAAGAGCTCACCCTGAGCTGC[C/T]ACCTTTTAATTTTTAATGCAATGTA
Phenotype
MIM: 600694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL6ST PubMed Links

Gene Details

Gene
IL6ST
Gene Name
interleukin 6 signal transducer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190981.1 6812 UTR 3 NP_001177910.1
NM_002184.3 6812 UTR 3 NP_002175.2
NM_175767.2 6812 UTR 3 NP_786943.1

View Full Product Details