Product Details

SNP ID
rs1657904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54360753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGCTTATATTGATGATTTTGAT[C/T]TAGGCCAAATATATCCTGGTGCAAG
Phenotype
MIM: 613258 MIM: 607051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C18orf54 PubMed Links

Gene Details

Gene
C18orf54
Gene Name
chromosome 18 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288980.1 315 Silent Mutation CTA,TTA L61L NP_001275909.1
NM_001288981.1 315 Silent Mutation CTA,TTA L61L NP_001275910.1
NM_001288982.1 315 Silent Mutation CTA,TTA L61L NP_001275911.1
NM_173529.5 315 Silent Mutation CTA,TTA L61L NP_775800.3
XM_005258201.1 315 Silent Mutation CTA,TTA L61L XP_005258258.1
XM_005258203.1 315 Silent Mutation CTA,TTA L61L XP_005258260.1
XM_017025567.1 315 Silent Mutation CTA,TTA L61L XP_016881056.1
Gene
STARD6
Gene Name
StAR related lipid transfer domain containing 6
There are no transcripts associated with this gene.

View Full Product Details