Product Details

SNP ID
rs1047108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75627097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGTGGGCCTCATCCTGACTTA[A/G]AACTCGGGGAGGGGCCACTCTTCCT
Phenotype
MIM: 603781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO15B PubMed Links

Gene Details

Gene
MYO15B
Gene Name
myosin XVB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309242.1 2405 Intron NP_001296171.1
XM_017025120.1 2405 Intron XP_016880609.1
XM_017025121.1 2405 Intron XP_016880610.1
XM_017025122.1 2405 Intron XP_016880611.1
XM_017025123.1 2405 Intron XP_016880612.1
XM_017025124.1 2405 Intron XP_016880613.1
XM_017025125.1 2405 Intron XP_016880614.1
XM_017025126.1 2405 Intron XP_016880615.1
XM_017025127.1 2405 Intron XP_016880616.1
XM_017025128.1 2405 Intron XP_016880617.1
XM_017025129.1 2405 Intron XP_016880618.1
XM_017025130.1 2405 Intron XP_016880619.1
XM_017025131.1 2405 Intron XP_016880620.1
XM_017025132.1 2405 Intron XP_016880621.1
XM_017025133.1 2405 Intron XP_016880622.1
XM_017025134.1 2405 Intron XP_016880623.1
XM_017025135.1 2405 Intron XP_016880624.1
XM_017025136.1 2405 Intron XP_016880625.1
XM_017025137.1 2405 Intron XP_016880626.1
XM_017025138.1 2405 Intron XP_016880627.1
XM_017025139.1 2405 Intron XP_016880628.1
XM_017025140.1 2405 Intron XP_016880629.1
XM_017025141.1 2405 Intron XP_016880630.1
XM_017025142.1 2405 Intron XP_016880631.1
XM_017025143.1 2405 Intron XP_016880632.1
XM_017025144.1 2405 Intron XP_016880633.1
Gene
RECQL5
Gene Name
RecQ like helicase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003715.3 2405 Intron NP_001003715.1
NM_001003716.3 2405 Intron NP_001003716.1
NM_004259.6 2405 UTR 3 NP_004250.4
XM_005257818.3 2405 UTR 3 XP_005257875.1
XM_005257822.3 2405 UTR 3 XP_005257879.1
XM_005257823.3 2405 Intron XP_005257880.1
XM_006722186.2 2405 UTR 3 XP_006722249.1
XM_011525482.2 2405 UTR 3 XP_011523784.1
XM_011525484.1 2405 UTR 3 XP_011523786.1
XM_011525485.2 2405 UTR 3 XP_011523787.1
XM_011525486.2 2405 UTR 3 XP_011523788.1
XM_017025343.1 2405 UTR 3 XP_016880832.1
XM_017025344.1 2405 Intron XP_016880833.1
Gene
SMIM5
Gene Name
small integral membrane protein 5
There are no transcripts associated with this gene.

View Full Product Details