Product Details
- SNP ID
-
rs829121
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:50843148 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGGAGGGAGGGAGGGGCGCATGCA[A/G]AGGGCAGTACCTGTTCGTGCCTGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMPRSS12
PubMed Links
Gene Details
- Gene
- TMPRSS12
- Gene Name
- transmembrane protease, serine 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182559.2 |
231 |
Missense Mutation |
AAG,GAG |
K62E |
NP_872365.1 |
View Full Product Details