Product Details
- SNP ID
-
rs1552473
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30253942 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGGGTAGGGTCAGTCTGTAAAG[A/G]GAAAAACAGTGTCTGTGAATATAGC
- Phenotype
-
MIM: 602403
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BLMH
PubMed Links
Gene Details
- Gene
- BLMH
- Gene Name
- bleomycin hydrolase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000386.3 |
|
Intron |
|
|
NP_000377.1 |
View Full Product Details