Product Details

SNP ID
rs1127742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32334591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGAATAAGTCCTTGAAGACTG[G/T]CCACACTCAGAAACCTGCCACACAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022344.3 837 Missense Mutation NP_071739.2
XM_005258022.3 837 Missense Mutation XP_005258079.1
XM_017024940.1 837 Missense Mutation XP_016880429.1
XM_017024941.1 837 Missense Mutation XP_016880430.1
Gene
RHBDL3
Gene Name
rhomboid like 3
There are no transcripts associated with this gene.

View Full Product Details