Product Details

SNP ID
rs1050096
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:23558170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTCCACACCTCCACGCTCTGAA[A/G]AGACCTTAGAACTTGGGGGTGGGGT
Phenotype
MIM: 600277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ID3 PubMed Links

Gene Details

Gene
ID3
Gene Name
inhibitor of DNA binding 3, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002167.4 999 UTR 3 NP_002158.3

View Full Product Details