Product Details
- SNP ID
-
rs1683560
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:134121777 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTCTAAGGAGATGCGAGTGGGAA[A/G]CTGGGTCGTTGAACTGACCCCCACC
- Phenotype
-
MIM: 601774
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MGAT5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75251578] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MGAT5
- Gene Name
- mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase
- Gene
- MIR3679
- Gene Name
- microRNA 3679
There are no transcripts associated with this gene.
View Full Product Details