Product Details

SNP ID
rs1468317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50384878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTGTTCTGGGAATGCTTCAAGA[C/T]ACATCTGACCCAAACACCTGCACAG
Phenotype
MIM: 610885 MIM: 614854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EME1 PubMed Links

Gene Details

Gene
EME1
Gene Name
essential meiotic structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
LRRC59
Gene Name
leucine rich repeat containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018509.3 Intron NP_060979.2

View Full Product Details