Product Details

SNP ID
rs818202
Assay Type
Functionally tested
NCBI dbSNP Submissions
69
Location
Chr.1:20590298 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGTTTGGAAAGTTTCAGAGCTC[A/G]AAATCACAGAGCTGGGATTGGAACC
Phenotype
MIM: 123920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDA PubMed Links

Gene Details

Gene
CDA
Gene Name
cytidine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001785.2 Intron NP_001776.1

View Full Product Details