Product Details
- SNP ID
-
rs589942
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:20589587 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCTGGGGTGCAGGCAGGGAGAG[C/G]GTTCGTGCTTTCCCAGCCTCGGATA
- Phenotype
-
MIM: 123920
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDA
PubMed Links
Gene Details
- Gene
- CDA
- Gene Name
- cytidine deaminase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001785.2 |
|
Intron |
|
|
NP_001776.1 |
View Full Product Details