Product Details

SNP ID
rs1396958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190205464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTAACTTCCTAAGCCTCATGT[C/T]TTAGCTTTAATAAAGCCTACTGAAT
Phenotype
MIM: 615117 MIM: 610690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf88 PubMed Links

Gene Details

Gene
C2orf88
Gene Name
chromosome 2 open reading frame 88
There are no transcripts associated with this gene.

Gene
HIBCH
Gene Name
3-hydroxyisobutyryl-CoA hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014362.3 Intron NP_055177.2
NM_198047.2 Intron NP_932164.1
XM_011510953.1 Intron XP_011509255.1
XM_011510954.1 Intron XP_011509256.1

View Full Product Details