Product Details

SNP ID
rs13796
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:154273441 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCGTGGGAACCCAAGGTTCCATAG[C/T]CCTCAGCACCCCCCTGAGGAATTTG
Phenotype
MIM: 605998 MIM: 616472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAX1 PubMed Links

Gene Details

Gene
HAX1
Gene Name
HCLS1 associated protein X-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018837.1 Intron NP_001018238.1
NM_006118.3 Intron NP_006109.2
Gene
UBAP2L
Gene Name
ubiquitin associated protein 2 like
There are no transcripts associated with this gene.

View Full Product Details