Product Details

SNP ID
rs1407128
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:160055888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCCCCCGTTCCGTGGATTCCA[C/T]GTCACCTCTGCCCTGGGTGTCCAAG
Phenotype
MIM: 602208 MIM: 600932
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNJ10 PubMed Links

Gene Details

Gene
KCNJ10
Gene Name
potassium voltage-gated channel subfamily J member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002241.4 888 Intron NP_002232.2
Gene
KCNJ9
Gene Name
potassium voltage-gated channel subfamily J member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004983.2 888 Intron NP_004974.2
XM_017001247.1 888 UTR 5 XP_016856736.1

View Full Product Details