Product Details

SNP ID
rs1047449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41393899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTGAGTTTCTTGGCTGCCTTAC[G/T]CGGGCAACTCCAGCCATGCAAATGA
Phenotype
MIM: 601077
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRT31 PubMed Links

Gene Details

Gene
KRT31
Gene Name
keratin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002277.2 1421 UTR 3 NP_002268.2
Gene
LOC100505782
Gene Name
uncharacterized LOC100505782
There are no transcripts associated with this gene.

View Full Product Details