Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164269.1 | 239 | Intron | NP_001157741.1 | ||
NM_001164270.1 | 239 | UTR 5 | NP_001157742.1 | ||
NM_002875.4 | 239 | UTR 5 | NP_002866.2 | ||
NM_133487.3 | 239 | UTR 5 | NP_597994.3 | ||
XM_006720626.3 | 239 | Intron | XP_006720689.1 | ||
XM_011521857.2 | 239 | Intron | XP_011520159.1 | ||
XM_011521858.2 | 239 | Intron | XP_011520160.1 | ||
XM_011521859.2 | 239 | Intron | XP_011520161.1 | ||
XM_011521860.2 | 239 | Intron | XP_011520162.1 | ||
XM_011521861.2 | 239 | Intron | XP_011520163.1 | ||
XM_011521862.2 | 239 | UTR 5 | XP_011520164.1 |